Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000346832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000355394.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000359188.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000376134.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000376138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000376139.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000376140.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000376142.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000376160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000376166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
ENST00000376170.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
NM_001012750.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
NM_001012751.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
NM_001012752.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
NM_001178116.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
NM_001178119.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
NM_001178120.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
NM_001178121.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
NM_005470.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10014008.hg.1 |
TC10001120.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000346832.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000359188.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376140.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376170.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001012750.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001012751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001012752.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001178116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001178119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001178120.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001178121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_005470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000346832.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000359188.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376138.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376139.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376140.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376142.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
ENST00000376170.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001012750.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001012751.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001012752.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001178116.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001178119.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001178120.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_001178121.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
NM_005470.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168746.hg.1 |
TC1000010101.hg.1 |
Clariom D TC |