Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000486782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000486782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000486782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000462082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000484727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000486782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000486782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000486782.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
ENST00000495216.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009016.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000286371.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000465172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000466678.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000475483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000482635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
ENST00000487199.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
NM_001679.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164694.hg.1 |
TC0300009016.hg.1 |
Clariom D TC |