Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000359314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000463175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000463175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000463175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000463175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000477159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000477159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000477159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000479857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000479857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000479857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000479857.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000486693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000486693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000486693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
ENST00000486693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008183.hg.1 |
NM_012120.2 |
Refseq |