Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044770.hg.1 |
TC01002870.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044770.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044770.hg.1 |
ENST00000401027.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044770.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044770.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044770.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044770.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044770.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044773.hg.1 |
TC01002870.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044773.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211262.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211262.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211262.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211262.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211262.hg.1 |
TC0100014937.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014937.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000370310.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000613047.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000613902.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000615519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
ENST00000616709.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001006605.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001252269.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001252270.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001252271.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
NM_001252273.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100211259.hg.1 |
TC0100014937.hg.1 |
Clariom D TC |