Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008672.hg.1 |
TC01000545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008672.hg.1 |
ENST00000310955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008672.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008672.hg.1 |
ENST00000437896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008672.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008673.hg.1 |
TC01000545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008673.hg.1 |
ENST00000310955.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008673.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008673.hg.1 |
ENST00000437896.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01008673.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000482046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000482046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000482046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
ENST00000482046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156894.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156894.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156894.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156894.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156894.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100156894.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100156894.hg.1 |
TC0100008057.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000310955.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000372462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000478882.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000482046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000482046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000482046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
ENST00000482046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008057.hg.1 |
NM_001255.2 |
Refseq |