Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001794.hg.1 |
TC6_mann_hap4000111.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001794.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001794.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002249.hg.1 |
TC6_qbl_hap6000123.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002249.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002249.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002274.hg.1 |
TC6_cox_hap2000133.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002274.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002274.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002204.hg.1 |
TC6_dbb_hap3000122.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002204.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002204.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002048.hg.1 |
TC6_ssto_hap7000114.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002048.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002048.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016825.hg.1 |
TC06001498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016825.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016825.hg.1 |
ENST00000376706.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016825.hg.1 |
ENST00000449742.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016825.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016825.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000932.hg.1 |
TC6_apd_hap1000069.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000932.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000932.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001995.hg.1 |
TC6_mcf_hap5000113.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001995.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001995.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011354.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176444.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176444.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176444.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176444.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176444.hg.1 |
ENST00000376704.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176444.hg.1 |
ENST00000449742.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176444.hg.1 |
NM_006778.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176444.hg.1 |
NM_052828.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600176444.hg.1 |
TC0600011354.hg.1 |
Clariom D TC |