Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145064.2 | NP_001138536.1 | 9 | 5126 | 112 | |
GenBank mRNA | AK123110.1 | - | 1 | 1971 | 112 | |
AL832384.1 | - | 1 | 2041 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |