Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019603.hg.1 |
ENST00000167588.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019603.hg.1 |
NM_019010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019603.hg.1 |
TC17001473.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000482529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000482529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010640.hg.1 |
ENST00000482529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000482529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000482529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010640.hg.1 |
ENST00000482529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184017.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184017.hg.1 |
ENST00000167588.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184017.hg.1 |
TC1700010640.hg.1 |
Clariom D TC |