Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009933.hg.1 |
TC09000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009933.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009933.hg.1 |
ENST00000495524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009933.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000486887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000486887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000486887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009934.hg.1 |
TC09000773.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009934.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009934.hg.1 |
ENST00000495524.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009934.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161746.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161746.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161746.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161746.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161746.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161746.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161746.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161746.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161746.hg.1 |
TC0900009074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900161745.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161745.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161745.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161745.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161745.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161745.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900161745.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161745.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900161745.hg.1 |
TC0900009074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000371964.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000486887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000486887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000486887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
ENST00000495524.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_001278928.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009074.hg.1 |
NM_017503.4 |
Refseq |