Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000352832.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000354983.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000358244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000359106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000360761.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000416767.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000429973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000442258.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000502264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000503485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000505165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000507336.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000507510.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000507609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000507896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000510115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000510366.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000512389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000513689.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000513964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000514079.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000514181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000514717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000515165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000515411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
ENST00000515765.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256324.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256326.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256327.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256328.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256329.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256330.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256332.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256333.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256334.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256359.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256360.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_001256361.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_018896.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198376.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198377.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198378.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198379.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198384.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198387.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198388.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NM_198396.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NR_046054.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NR_046055.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NR_046056.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NR_046057.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
NR_046058.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17008690.hg.1 |
TC17000671.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000352832.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000354983.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000358244.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000359106.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000360761.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000416767.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000429973.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000442258.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000502264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000503436.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000503485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000503607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000504076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000505165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000506406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000507336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000507510.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000507609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000507896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000510115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000510366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000511765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000511768.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000512389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000513689.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000513964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000514079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000514181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000514717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000515165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000515411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000515765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256326.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256330.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256359.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256360.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_018896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198378.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046056.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046057.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000352832.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000354983.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000358244.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000359106.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000360761.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000416767.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000429973.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000442258.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000502264.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000503436.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000503485.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000503607.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000504076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000505165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000506406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000507336.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000507510.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000507609.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000507896.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000510115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000510366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000511765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000511768.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000512389.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000513689.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000513964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000514079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000514181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000514717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000515165.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000515411.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
ENST00000515765.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256324.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256326.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256327.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256328.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256329.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256330.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256332.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256333.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256334.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256359.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256360.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_001256361.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_018896.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198376.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198377.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198378.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198379.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198384.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198387.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198388.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NM_198396.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046054.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046055.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046056.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046057.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
NR_046058.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700161460.hg.1 |
TC1700008262.hg.1 |
Clariom D TC |