Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019954.hg.1 |
TC06001675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019954.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019954.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019955.hg.1 |
TC06001675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019955.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06019955.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011661.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180013.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180013.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180013.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180013.hg.1 |
ENST00000229812.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600180013.hg.1 |
NM_001305102.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180013.hg.1 |
NM_007271.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600180013.hg.1 |
TC0600011661.hg.1 |
Clariom D TC |