Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000482552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000482552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000482552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
ENST00000482552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006678.hg.1 |
TC09000585.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006678.hg.1 |
ENST00000288502.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006678.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006678.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006677.hg.1 |
TC09000585.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006677.hg.1 |
ENST00000288502.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006677.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006677.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006679.hg.1 |
TC09000585.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006679.hg.1 |
ENST00000288502.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006679.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006679.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000471206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000482552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000482552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000482552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
ENST00000482552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008516.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157020.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157020.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157020.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157020.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157020.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157020.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157020.hg.1 |
TC0900008516.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900157015.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157015.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157015.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157015.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157015.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157015.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157015.hg.1 |
TC0900008516.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900157016.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157016.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157016.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157016.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157016.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157016.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157016.hg.1 |
TC0900008516.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900157018.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157018.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157018.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157018.hg.1 |
ENST00000288502.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157018.hg.1 |
ENST00000374049.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900157018.hg.1 |
NM_153045.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900157018.hg.1 |
TC0900008516.hg.1 |
Clariom D TC |