Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001873.hg.1 |
TC03000088.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001873.hg.1 |
ENST00000285042.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001873.hg.1 |
ENST00000303688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001873.hg.1 |
ENST00000383794.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001873.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000476763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000476763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000476763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000476763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001872.hg.1 |
TC03000088.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001872.hg.1 |
ENST00000285042.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001872.hg.1 |
ENST00000303688.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001872.hg.1 |
ENST00000383794.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03001872.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146604.hg.1 |
TC0300006687.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146602.hg.1 |
TC0300006687.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000303688.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000383794.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000463438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000465759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000476763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000476763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000476763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000476763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
ENST00000611584.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006687.hg.1 |
NM_016474.4 |
Refseq |