Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
ENST00000477973.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011525.hg.1 |
NM_012234.6 |
Refseq |