Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016418.5 | NP_057502.2 | 16 - 17 | 2227 | 74 | |
NM_181828.2 | NP_861966.1 | 15 - 16 | 2101 | 74 | ||
NM_181829.2 | NP_861967.1 | 15 - 16 | 2104 | 74 | ||
NM_181830.2 | NP_861968.1 | 14 - 15 | 1978 | 74 | ||
XM_017028810.1 | XP_016884299.1 | 16 - 17 | 1786 | 74 | ||
GenBank mRNA | AF369658.1 | - | 16 - 17 | 2209 | 74 | |
AF369663.1 | - | 15 - 16 | 1658 | 74 | ||
AF369664.1 | - | 15 - 16 | 1661 | 74 | ||
AF369665.1 | - | 14 - 15 | 1535 | 74 | ||
AF369666.1 | - | 17 - 18 | 1900 | 74 | ||
AF369667.1 | - | 16 - 17 | 1809 | 74 | ||
AK297116.1 | - | 16 - 17 | 2132 | 74 | ||
S73854.1 | - | 2 - 3 | 65 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |