Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005560.4 | NP_005551.3 | 46 - 47 | 6235 | 99 | |
XM_006723796.3 | XP_006723859.1 | 46 - 47 | 6238 | 99 | ||
XM_006723798.3 | XP_006723861.1 | 46 - 47 | 6241 | 99 | ||
XM_011528818.2 | XP_011527120.1 | 45 - 46 | 6100 | 99 | ||
XM_011528819.2 | XP_011527121.1 | 44 - 45 | 5965 | 99 | ||
GenBank mRNA | AF443072.1 | - | 46 - 47 | 6235 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |