Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004169.hg.1 |
ENST00000243903.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004169.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20004169.hg.1 |
TC20000289.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007329.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150774.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150774.hg.1 |
NM_024855.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000150774.hg.1 |
TC2000007329.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000150774.hg.1 |
ENST00000243903.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000150774.hg.1 |
NM_024855.3 |
Refseq |