Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008398.hg.1 |
TC09000691.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008398.hg.1 |
ENST00000303890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008398.hg.1 |
ENST00000351030.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008398.hg.1 |
ENST00000448249.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008398.hg.1 |
ENST00000535026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008398.hg.1 |
NM_001242352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008398.hg.1 |
NM_001242354.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008398.hg.1 |
NM_002540.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09008398.hg.1 |
NM_153435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000351030.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000372807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000393527.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000444119.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000444993.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000448249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000469582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
NM_001242352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
NM_001242354.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
NM_002540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
NM_153435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000351030.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000372807.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000393527.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000444119.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000444993.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000448249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
ENST00000469582.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
NM_001242352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
NM_001242354.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
NM_002540.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
NM_153435.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900159561.hg.1 |
TC0900008869.hg.1 |
Clariom D TC |