Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
ENST00000483821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000481441.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
ENST00000483821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011525.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
ENST00000483821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
ENST00000374680.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
ENST00000374685.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
ENST00000483281.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
ENST00000483821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
NM_001291989.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
NM_021976.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600178842.hg.1 |
TC0600011525.hg.1 |
Clariom D TC |