Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002429.hg.1 |
ENST00000215743.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002429.hg.1 |
ENST00000477567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002429.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002429.hg.1 |
TC22000138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000480185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000480185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000480185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000480185.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000492464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000492464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000492464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000492464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002426.hg.1 |
ENST00000215743.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002426.hg.1 |
ENST00000477567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002426.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002426.hg.1 |
TC22000138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002427.hg.1 |
ENST00000215743.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002427.hg.1 |
ENST00000477567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002427.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22002427.hg.1 |
TC22000138.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
TC2200009229.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167801.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000428253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000434318.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000465730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000480185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000480185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000480185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000480185.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000488363.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000492464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000492464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000492464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000492464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
ENST00000493132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009229.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167799.hg.1 |
TC2200009229.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200167799.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167799.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167799.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167799.hg.1 |
NM_005940.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167799.hg.1 |
ENST00000215743.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167799.hg.1 |
ENST00000437086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167799.hg.1 |
ENST00000460352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167799.hg.1 |
NM_005940.4 |
Refseq |