Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321851.1 | NP_001308780.1 | 8 - 9 | 1601 | 140 | |
NM_004239.4 | NP_004230.2 | 8 - 9 | 1604 | 140 | ||
XM_017021787.1 | XP_016877276.1 | 9 - 10 | 1625 | 140 | ||
GenBank mRNA | AF007217.1 | - | 8 - 9 | 1581 | 140 | |
AK307489.1 | - | 8 - 9 | 1604 | 140 | ||
BC146845.1 | - | 8 - 9 | 1486 | 9056773 | 140 | |
Y12490.1 | - | 8 - 9 | 1586 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |