Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541800.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541800.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541800.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
ENST00000541800.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016523.hg.1 |
ENST00000240617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016523.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016523.hg.1 |
TC12001269.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541800.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541800.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541800.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
ENST00000541800.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010006.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175542.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175542.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175542.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175542.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175542.hg.1 |
ENST00000240617.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175542.hg.1 |
ENST00000540572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175542.hg.1 |
ENST00000541618.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175542.hg.1 |
NM_024829.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175542.hg.1 |
TC1200010006.hg.1 |
Clariom D TC |