Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002044.hg.1 |
ENST00000261292.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002044.hg.1 |
ENST00000427224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002044.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002044.hg.1 |
TC18000175.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002043.hg.1 |
ENST00000261292.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002043.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002043.hg.1 |
TC18000175.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000583083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000583083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000583083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000623277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000623277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
ENST00000623277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800149948.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149948.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149948.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149948.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800149948.hg.1 |
TC1800007298.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800149948.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149948.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149948.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149948.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000261292.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000427224.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000577628.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000579750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000580036.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000583083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000583083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000583083.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000623277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000623277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
ENST00000623277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_001308006.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007298.hg.1 |
NM_006033.3 |
Refseq |