Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023281.hg.1 |
TC04001670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023281.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023281.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023279.hg.1 |
TC04001670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023279.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023279.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023280.hg.1 |
TC04001670.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023280.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04023280.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183890.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183890.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183890.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183890.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183890.hg.1 |
TC0400012213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400183887.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183887.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183887.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183887.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183887.hg.1 |
TC0400012213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012213.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183888.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183888.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183888.hg.1 |
ENST00000433477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400183888.hg.1 |
NM_001334.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400183888.hg.1 |
TC0400012213.hg.1 |
Clariom D TC |