Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004753.hg.1 |
ENST00000308748.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004753.hg.1 |
ENST00000414952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004753.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004753.hg.1 |
TC16000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004757.hg.1 |
ENST00000308748.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004757.hg.1 |
ENST00000414952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004757.hg.1 |
ENST00000483405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004757.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004757.hg.1 |
TC16000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004755.hg.1 |
ENST00000308748.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004755.hg.1 |
ENST00000414952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004755.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004755.hg.1 |
TC16000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004756.hg.1 |
ENST00000308748.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004756.hg.1 |
ENST00000414952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004756.hg.1 |
ENST00000483405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004756.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004756.hg.1 |
TC16000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004751.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004751.hg.1 |
TC16000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004752.hg.1 |
ENST00000414952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004752.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004752.hg.1 |
TC16000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007431.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153871.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153871.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153871.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153871.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153871.hg.1 |
TC1600007431.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600153872.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153872.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153872.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153872.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153872.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153872.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153872.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153872.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153872.hg.1 |
TC1600007431.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600153873.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153873.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153873.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153873.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153873.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153873.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153873.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153873.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153873.hg.1 |
TC1600007431.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153874.hg.1 |
TC1600007431.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000308748.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000414952.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000483405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000563177.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
ENST00000566693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
NM_181718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600153875.hg.1 |
TC1600007431.hg.1 |
Clariom D TC |