Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRIQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079910.1 | NP_001073379.1 | 14 - 15 | 3384 | 123 | |
XM_011538817.2 | XP_011537119.1 | 14 - 15 | 3327 | 123 | ||
XM_011538818.2 | XP_011537120.1 | 14 - 15 | 3327 | 123 | ||
XM_011538819.2 | XP_011537121.1 | 14 - 15 | 3327 | 123 | ||
XM_011538823.2 | XP_011537125.1 | 14 - 15 | 3327 | 123 | ||
XM_011538824.2 | XP_011537126.1 | 14 - 15 | 3327 | 123 | ||
XM_017020019.1 | XP_016875508.1 | 14 - 15 | 3327 | 123 | ||
GenBank mRNA | AB058704.1 | - | 10 - 11 | 2948 | 123 | |
AK307799.1 | - | 14 - 15 | 3335 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |