Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017651.hg.1 |
ENST00000328194.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017651.hg.1 |
ENST00000530044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017651.hg.1 |
NM_001243254.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017651.hg.1 |
NM_015213.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017651.hg.1 |
TC11001397.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
ENST00000328194.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
ENST00000526707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
ENST00000530044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
ENST00000530780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
NM_001243254.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
NM_015213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
ENST00000328194.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
ENST00000526707.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
ENST00000530044.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
ENST00000530780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
NM_001243254.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
NM_015213.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176281.hg.1 |
TC1100010058.hg.1 |
Clariom D TC |