Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019405.hg.1 |
ENST00000298992.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019405.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019405.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11019405.hg.1 |
TC11001544.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000530814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000530814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000530814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
ENST00000530814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179713.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179713.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179713.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100179713.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100179713.hg.1 |
TC1100010505.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000435224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000530814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000530814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000530814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
ENST00000530814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010505.hg.1 |
NM_145804.2 |
Refseq |