Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008411.hg.1 |
ENST00000342812.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008411.hg.1 |
ENST00000377666.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008411.hg.1 |
ENST00000377669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008411.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008411.hg.1 |
TC13000733.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300162116.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162116.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162116.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162116.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162116.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300162116.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300162116.hg.1 |
TC1300009249.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000377669.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
ENST00000472022.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009249.hg.1 |
NM_007249.4 |
Refseq |