Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003804.hg.1 |
ENST00000220478.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003804.hg.1 |
ENST00000542355.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003804.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003804.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003804.hg.1 |
TC15000409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003803.hg.1 |
ENST00000220478.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003803.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003803.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003803.hg.1 |
TC15000409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003806.hg.1 |
ENST00000220478.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003806.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003806.hg.1 |
TC15000409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003805.hg.1 |
ENST00000220478.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003805.hg.1 |
ENST00000542355.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003805.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003805.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003805.hg.1 |
TC15000409.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500151600.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151600.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151600.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151600.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151600.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151600.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151600.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151600.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151600.hg.1 |
TC1500007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151601.hg.1 |
TC1500007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007237.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151602.hg.1 |
TC1500007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
ENST00000542355.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
ENST00000558709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151603.hg.1 |
TC1500007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500151604.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151604.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151604.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151604.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151604.hg.1 |
TC1500007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500151599.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151599.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151599.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151599.hg.1 |
ENST00000220478.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151599.hg.1 |
NM_001165257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151599.hg.1 |
NM_013243.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151599.hg.1 |
TC1500007237.hg.1 |
Clariom D TC |