Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000050.hg.1 |
ENST00000327158.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000050.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000050.hg.1 |
TC17000007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006480.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700144297.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144297.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700144297.hg.1 |
ENST00000327158.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144297.hg.1 |
NM_013337.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700144297.hg.1 |
TC1700006480.hg.1 |
Clariom D TC |