Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193376.1 | NP_001180305.1 | 12 - 13 | 2902 | 71 | |
NM_198253.2 | NP_937983.2 | 13 - 14 | 3091 | 71 | ||
XM_017009796.1 | XP_016865285.1 | 13 - 14 | 3663 | 71 | ||
GenBank mRNA | AB085628.1 | - | 12 - 13 | 2844 | 71 | |
AB086379.1 | - | 10 - 11 | 2662 | 71 | ||
AB086950.1 | - | 10 - 11 | 2626 | 71 | ||
AF015950.1 | - | 13 - 14 | 3088 | 71 | ||
AF018167.1 | - | 13 - 14 | 3089 | 71 | ||
AX810036.1 | - | 13 - 14 | 3088 | 71 | ||
AX810378.1 | - | 13 - 14 | 3088 | 71 | ||
BC062321.1 | - | 9 - 10 | 1664 | 71 | ||
JF896281.1 | - | 2 - 3 | 125 | 71 | ||
JF896283.1 | - | 6 - 7 | 703 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |