Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003355.hg.1 |
ENST00000261596.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003355.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003355.hg.1 |
TC18000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000581568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000581568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000581568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000584915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000584915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000584915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
ENST00000584915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800154073.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154073.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800154073.hg.1 |
TC1800007905.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800154073.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154073.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000261596.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000581568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000581568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000581568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000584294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000584915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000584915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000584915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
ENST00000584915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007905.hg.1 |
NM_014646.2 |
Refseq |