Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172173.2 | NP_751913.1 | 14 - 15 | 1149 | 69 | |
XM_005270198.1 | XP_005270255.1 | 15 - 16 | 1203 | 69 | ||
XM_005270201.1 | XP_005270258.1 | 14 - 15 | 1140 | 69 | ||
XM_017016722.1 | XP_016872211.1 | 15 - 16 | 1203 | 69 | ||
XM_017016726.1 | XP_016872215.1 | 14 - 15 | 1140 | 69 | ||
GenBank mRNA | U32472.1 | - | 1 - 2 | 44 | 69 | |
U50359.1 | - | 3 - 4 | 116 | 69 | ||
U50360.1 | - | 3 - 4 | 116 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Protein Phosphatase Homo sapiens Probe spans exons |