Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
TC04000254.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
ENST00000313860.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
ENST00000503057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
ENST00000508501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
ENST00000509638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
ENST00000511496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
ENST00000512632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
ENST00000512820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
ENST00000512946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
ENST00000513024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
NM_001112717.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
NM_001112718.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003693.hg.1 |
NM_014988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000313860.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000446625.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000503057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000508501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000509638.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000511424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000511496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000512632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000512820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000512946.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000513024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000514190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_001112717.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_001112718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_001289122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_001289124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_014988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000313860.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000446625.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000503057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000508501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000509638.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000511424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000511496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000512632.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000512820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000512946.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000513024.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
ENST00000514190.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_001112717.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_001112718.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_001289122.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_001289124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
NM_014988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400150213.hg.1 |
TC0400007345.hg.1 |
Clariom D TC |