Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252195.1 | NP_001239124.1 | 3 - 4 | 381 | 108 | |
XM_005251616.4 | XP_005251673.1 | 5 - 6 | 3902 | 108 | ||
XM_005251618.4 | XP_005251675.1 | 3 - 4 | 449 | 108 | ||
XM_011518055.2 | XP_011516357.1 | 3 - 4 | 723 | 108 | ||
GenBank mRNA | BI561111.1 | - | 3 - 4 | 346 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |