Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024305.hg.1 |
TC04001762.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024305.hg.1 |
ENST00000296525.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024305.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024305.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024306.hg.1 |
TC04001762.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024306.hg.1 |
ENST00000296525.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024306.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04024306.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000505299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000505299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000505299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000505299.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000510578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000510578.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000511879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000511879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000511879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000511879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185572.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185572.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185572.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185572.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185572.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185572.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185572.hg.1 |
TC0400012478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000505299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000505299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000505299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000505299.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000510578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000510578.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000511879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000511879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000511879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000511879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400012478.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185573.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185573.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185573.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185573.hg.1 |
ENST00000296525.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185573.hg.1 |
ENST00000512254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400185573.hg.1 |
NM_080874.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400185573.hg.1 |
TC0400012478.hg.1 |
Clariom D TC |