Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011731.hg.1 |
ENST00000304613.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011731.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011731.hg.1 |
ENST00000368572.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011731.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10011731.hg.1 |
TC10000949.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000478074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000478074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000478074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000478074.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000485110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000485110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000485110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000485110.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000530127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000530127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000530127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
ENST00000530127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000478074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000478074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000478074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000478074.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000485110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000485110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000485110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000485110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000530127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000530127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000530127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
ENST00000530127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009447.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000164555.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000164555.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000164555.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000164555.hg.1 |
ENST00000304613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000164555.hg.1 |
ENST00000368571.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000164555.hg.1 |
NM_152643.7 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000164555.hg.1 |
TC1000009447.hg.1 |
Clariom D TC |