Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048436.hg.1 |
TC01003124.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048436.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048436.hg.1 |
ENST00000425272.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048436.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048434.hg.1 |
TC01003124.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048434.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048434.hg.1 |
ENST00000425272.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048434.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01048434.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216962.hg.1 |
TC0100015627.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
ENST00000496858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015627.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
ENST00000254101.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
ENST00000474939.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
NM_005399.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
NR_103870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
NR_103871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100216964.hg.1 |
TC0100015627.hg.1 |
Clariom D TC |