Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036778.hg.1 |
TC03002077.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036778.hg.1 |
ENST00000421852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036778.hg.1 |
ENST00000475480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03036778.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
ENST00000467520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000296254.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
ENST00000467520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300014081.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209661.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209661.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209661.hg.1 |
ENST00000467520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209661.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209661.hg.1 |
ENST00000421852.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209661.hg.1 |
ENST00000467061.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209661.hg.1 |
ENST00000467520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300209661.hg.1 |
NM_080652.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300209661.hg.1 |
TC0300014081.hg.1 |
Clariom D TC |