Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099952.2 | NP_001093422.2 | 46 - 47 | 6595 | 94 | |
NM_001168272.1 | NP_001161744.1 | 48 - 49 | 6694 | 94 | ||
NM_002222.5 | NP_002213.5 | 45 - 46 | 6550 | 94 | ||
XM_005265109.2 | XP_005265166.1 | 47 - 48 | 6670 | 94 | ||
XM_005265110.2 | XP_005265167.1 | 46 - 47 | 6622 | 94 | ||
XM_006713131.2 | XP_006713194.1 | 47 - 48 | 6673 | 94 | ||
XM_011533681.1 | XP_011531983.1 | 49 - 50 | 6742 | 94 | ||
XM_011533682.2 | XP_011531984.1 | 49 - 50 | 6668 | 94 | ||
XM_011533683.2 | XP_011531985.1 | 49 - 50 | 6739 | 94 | ||
XM_011533684.1 | XP_011531986.1 | 49 - 50 | 6715 | 94 | ||
XM_011533685.1 | XP_011531987.1 | 48 - 49 | 6709 | 94 | ||
XM_011533686.1 | XP_011531988.1 | 48 - 49 | 6706 | 94 | ||
XM_011533687.1 | XP_011531989.1 | 48 - 49 | 6697 | 94 | ||
XM_011533688.1 | XP_011531990.1 | 48 - 49 | 6670 | 94 | ||
XM_011533690.1 | XP_011531992.1 | 49 - 50 | 6742 | 94 | ||
XM_017006357.1 | XP_016861846.1 | 49 - 50 | 6739 | 94 | ||
GenBank mRNA | AB208868.1 | - | 26 - 27 | 3466 | 94 | |
D26070.1 | - | 45 - 46 | 6456 | 94 | ||
L38019.2 | - | 46 - 47 | 6492 | 94 | ||
U23850.1 | - | 46 - 47 | 6344 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Homo sapiens Probe spans exons |