Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000488983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000488983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000488983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000488983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023414.hg.1 |
TC01001513.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023414.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023414.hg.1 |
ENST00000546011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023414.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000479505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000483154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000488983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000488983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000488983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000488983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
ENST00000493170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010654.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178341.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178341.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178341.hg.1 |
ENST00000209884.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178341.hg.1 |
NM_014458.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178341.hg.1 |
TC0100010654.hg.1 |
Clariom D TC |