Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044055.hg.1 |
TC01002831.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044055.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044055.hg.1 |
ENST00000431532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044055.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044055.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044057.hg.1 |
TC01002831.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044057.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044057.hg.1 |
ENST00000431532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044057.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044057.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044059.hg.1 |
TC01002831.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044059.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044059.hg.1 |
ENST00000431532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044059.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044059.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044052.hg.1 |
TC01002831.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044052.hg.1 |
ENST00000431532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044052.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044052.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044054.hg.1 |
TC01002831.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044054.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044054.hg.1 |
ENST00000431532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044054.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01044054.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210134.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210134.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210134.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210134.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210134.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210134.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210134.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210134.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210134.hg.1 |
TC0100014788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100210136.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210136.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210136.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210136.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210136.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210136.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210136.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210136.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210136.hg.1 |
TC0100014788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014788.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210132.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210132.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210132.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210132.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210132.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210132.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210132.hg.1 |
TC0100014788.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100210135.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210135.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210135.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210135.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210135.hg.1 |
ENST00000370574.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210135.hg.1 |
ENST00000431532.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100210135.hg.1 |
NM_001170670.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210135.hg.1 |
NM_017953.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100210135.hg.1 |
TC0100014788.hg.1 |
Clariom D TC |