Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010114.hg.1 |
TC02000646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010114.hg.1 |
ENST00000258457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010114.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010114.hg.1 |
ENST00000437250.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010114.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010115.hg.1 |
TC02000646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010115.hg.1 |
ENST00000258457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010115.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010115.hg.1 |
ENST00000437250.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010115.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010113.hg.1 |
TC02000646.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010113.hg.1 |
ENST00000258457.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010113.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010113.hg.1 |
ENST00000437250.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010113.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160989.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160989.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160989.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160989.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160989.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160989.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160989.hg.1 |
TC0200008734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200160991.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160991.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160991.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160991.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160991.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160991.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160991.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160991.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160991.hg.1 |
TC0200008734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008734.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160992.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160992.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160992.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160992.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160992.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160992.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160992.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160992.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160992.hg.1 |
TC0200008734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200160993.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160993.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160993.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160993.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160993.hg.1 |
ENST00000258457.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160993.hg.1 |
ENST00000410049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160993.hg.1 |
NM_001286537.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160993.hg.1 |
NM_024093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160993.hg.1 |
TC0200008734.hg.1 |
Clariom D TC |