Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021005.hg.1 |
TC09001650.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021005.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021005.hg.1 |
ENST00000450050.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021005.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09021005.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000277459.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011705.hg.1 |
NM_177999.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180067.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180067.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180067.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180067.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180067.hg.1 |
ENST00000277458.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180067.hg.1 |
ENST00000450050.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900180067.hg.1 |
NM_001202403.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180067.hg.1 |
NM_017873.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900180067.hg.1 |
TC0900011705.hg.1 |
Clariom D TC |