Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQSEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014869.6 | NP_055684.3 | 12 - 13 | 3044 | 139 | |
GenBank mRNA | AB018306.1 | - | 11 - 12 | 2537 | 139 | |
AY653734.1 | - | 12 - 13 | 2806 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |