Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019059.hg.1 |
TC09001520.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019059.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019059.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011318.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011318.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011318.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019060.hg.1 |
TC09001520.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019060.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019060.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177186.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177186.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177186.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177186.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177186.hg.1 |
TC0900011318.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900177188.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177188.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177188.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900177188.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900177188.hg.1 |
TC0900011318.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011318.hg.1 |
ENST00000374014.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011318.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011318.hg.1 |
NR_024032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011318.hg.1 |
NR_024032.1 |
Refseq |