Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002032.hg.1 |
TC03000092.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002032.hg.1 |
ENST00000253693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03002032.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146797.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146797.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146797.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146797.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146797.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146797.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146797.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300146797.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300146797.hg.1 |
TC0300006702.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000253693.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000415565.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000418994.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000443567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000457023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000463417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
ENST00000472400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006702.hg.1 |
NM_014296.2 |
Refseq |