Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000492828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
ENST00000492828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205619.hg.1 |
TC0300013888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205618.hg.1 |
TC0300013888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000492828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
ENST00000492828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013888.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205616.hg.1 |
TC0300013888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
ENST00000480589.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
ENST00000485421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205617.hg.1 |
TC0300013888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
ENST00000460316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205620.hg.1 |
TC0300013888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300205622.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205622.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205622.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205622.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205622.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205622.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205622.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205622.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205622.hg.1 |
TC0300013888.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300205621.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205621.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205621.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205621.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205621.hg.1 |
ENST00000466814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205621.hg.1 |
NM_001303264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205621.hg.1 |
NM_014779.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205621.hg.1 |
NR_130136.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205621.hg.1 |
TC0300013888.hg.1 |
Clariom D TC |