Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000262291.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000362134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000585847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000585847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000585847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000585847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000585847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000585847.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000586245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000586245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000586245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000586245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000586245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000586245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000586245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000586245.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587259.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587470.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000587945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588131.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588617.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000588915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000589823.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000589823.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000589823.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000589823.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000589823.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000589823.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000589823.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000589823.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000589823.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591782.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000591877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592106.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000592790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000593168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000593168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000593168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000593168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000593168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000593168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000593168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
ENST00000593168.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NM_030938.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008467.hg.1 |
NR_029493.1 |
Refseq |